missing translation for 'onlineSavingsMsg'
Learn More

Hyamine Hydroxide, Fisher BioReagents

Product Code. 11483094
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11483094 500 mL Amber Glass 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11483094 Supplier Fisher BioReagents Supplier No. BP2658500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 26248-39-5
Melting Point -30°C
Color Colorless
Density 0.933g/cm³
Boiling Point 65°C
Packaging Amber Glass
Molecular Formula C28H45NO3
Quantity 500 mL
Synonym hyamine hydroxide, methylbenzethonium hydroxide, benzyldimethyl 2-2-4-1,1,3,3-tetramethylbutyl tolyloxy ethoxy ethyl ammonium hydroxide, benzenemethanaminium, n,n-dimethyl-n-2-2-methyl-4-1,1,3,3-tetramethylbutyl phenoxy ethoxy ethyl-, hydroxide, n-benzyl-n,n-dimethyl-2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethanaminium hydroxide, benzenemethanaminium,n,n-dimethyl-n-2-2-methyl-4-1,1,3,3-tetramethylbutyl phenoxy ethoxy ethyl-,hydroxide 9ci, benzyl-dimethyl-2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium hydroxide, benzyldimethyl 2-2-2-methyl-4-2,4,4-trimethylpentan-2-yl phenoxy ethoxy ethyl azanium hydroxide
InChI Key MJAMCTLGWXIKOT-UHFFFAOYSA-M
SMILES CC1=C(C=CC(=C1)C(C)(C)CC(C)(C)C)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[OH-]
IUPAC Name benzyl-dimethyl-[2-[2-[2-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;hydroxide
Molecular Weight (g/mol) 443.672
PubChem CID 168497
Formula Weight 443.7
Physical Form Liquid
Chemical Name or Material Hyamine Hydroxide™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.