missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ HybridRack™ Pipet Tip, Extended Length
GREENER_CHOICE

Product Code. 30090454 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
10 μL
300 μL
1000 μL
Color:
Blue
Green
Red
Unit Size:
Pack of 960
This item is not returnable. View return policy

Product Code. 30090454

Brand: Axygen™ ERF300XTRS

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.

The Axygen HybridRack System combines sustainability and functionality to create a new, ecofriendly pipet tip product that is backed by Corning’s quality and value.

  • The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.
  • The plastic deck combined with the paperboard base provides a stable pipetting platform. The HybridRack is designed for end users to pipet directly from it, even when using a multi-channel instrument.
  • The paperboard base can be easily recycled through most local curbside recycling programs.
  • All HybridRack part numbers are provided sterile. Filtered, Maxymum Recovery, and non-filtered configurations are available.
  • The HybridRack System is manufactured in the USA at an ISO 13485, ISO 14001, and EnergyStar registered facility.
  • The pipet tips included in the HybridRack offering are the same tips as the Axygen MultiRack product line.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 300 μL
Filtered Filtered
Sterility Sterile
Color Green
Autoclavable Not Autoclavable
Format Rack
Tip Style Extended Length
Product Type Pipet Tip
Color Code Green
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.