missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ 0.1 N NaOH, Cytiva

Product Code. 17877707
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
50 L
100 L
200 L
Packaging:
Single-use Bioprocessing Container
Unit Size:
1 L
10 L
100 L
20 L
200 L
50 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
17877707 200 L Single-use Bioprocessing Container 200 L
17235886 1 L Single-use Bioprocessing Container 1 L
17245886 10 L Single-use Bioprocessing Container 10 L
17255886 20 L Single-use Bioprocessing Container 20 L
17847697 50 L Single-use Bioprocessing Container 50 L
17857677 100 L Single-use Bioprocessing Container 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17877707 Supplier HyClone products Cytiva Supplier No. SH30883.07
  • ONLINE EXCLUSIVE PRICE
    1550.00 €
    1475.00 € / 200 L
    Save 75.00 €
    4% Off
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Concentrated sodium hydroxide solutions for bioprocessing applications including pH adjustment and chromatography resin care.

HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean chromatography resins.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.1 μm sterile filtered.
  • All unit sizes are filled in single-use, ready-to-use bags.
  • Raw materials are USP/NF and EP compendial grade.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

Color Clear, Colorless
Concentration 0.1 M (0.1 N)
Quantity 200 L
Packaging Single-use Bioprocessing Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.