missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ 2 M NaCl, Cytiva

Product Code. 17837707
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
50 L
100 L
200 L
Packaging:
Single-use Bioprocessing Container
Unit Size:
1 L
10 L
100 L
20 L
200 L
50 L
Product Code. Quantity Packaging unitSize
17837707 50 L Single-use Bioprocessing Container 50 L
17285906 1 L Single-use Bioprocessing Container 1 L
17295906 10 L Single-use Bioprocessing Container 10 L
17205916 20 L Single-use Bioprocessing Container 20 L
17887687 100 L Single-use Bioprocessing Container 100 L
17867707 200 L Single-use Bioprocessing Container 200 L
6 options
This item is not returnable. View return policy

Product Code. 17837707

Brand: HyClone products Cytiva SH31141.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Concentrated sodium chloride solutions for applications in upstream and downstream bioprocessing.

HyClone™ NaCl solutions in single-use bags offer a convenient format for many upstream and downstream bioprocessing applications.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.2 μm sterile filtered.
  • Provided in single-use bag containers.
  • Raw material is EP, USP, and JP grade.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Clear, Colorless
Quantity 50 L
Concentration or Composition (by Analyte or Components) 2 M
Packaging Single-use Bioprocessing Container
Product Line HyClone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.