missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Alpha Calf Fraction Media Supplement, U.S. Origin

Product Code. 16615755
Change view
Click to view available options
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
16615755 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16615755 Supplier Cytiva Supplier No. SH30212.04

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Produced from bovine calf serum using a proprietary non-ethanol process to reduce immunoglobulins and proteins to levels similar to those of FBS

  • Optimized for the growth of hybridoma cells
  • Cost-effective medium supplement
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hybridoma
Country of Origin United States
Product Type Cell Culture Media Supplement
Sterilization Method Sequential Triple-filtered, 100 nm
Endotoxin Concentration ≤ 10 EU/mL
Hemoglobin Concentration ≤ 20 mg/dL
IgG levels ≤ 200 μg/mL
Iron Supplemented No
Product Line HyClone
Osmolality 265 to 310 mOsm/kg
Protein Content 3.2 to 4.7 gm/dL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.