missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Bovine Calf Serum, US Origin
Description
- Collected by venipuncture from formula-fed veal calves
- Age of animal at time of collection approximately 16-24 weeks
- Complete traceability back to original source
- Low in antibodies and high in growth factors
- Virus panel testing according to 9 CFR 113.53
- Sourced in the USA
Specifications
Specifications
| Certifications/Compliance | Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities |
| Content And Storage | ≤-10°C |
| Age | 16 to 22 weeks |
| Country of Origin | United States |
| Quantity | 50 mL |
| Sterilization Method | Sequential-filtered, 100 nm |
| Source | Formula-Fed Veal Calves |
| Tested For | Virus Panel Testing According to 9 CFR 113.53 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?