missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Bovine Calf Serum, US Origin

Product Code. 16675735
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16675735 500 mL 500mL
16655735 50 mL 50mL
16665735 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16675735 Supplier Cytiva Supplier No. SH30072.03NI

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A cost-effective alternative to Fetal Bovine Serum (FBS)

  • Collected by venipuncture from formula-fed veal calves
  • Age of animal at time of collection approximately 16-24 weeks
  • Complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
  • Sourced in the USA
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ≤-10°C
Age 16 to 22 weeks
Country of Origin United States
Quantity 500 mL
Sterilization Method Sequential-filtered, 100 nm
Source Formula-Fed Veal Calves
Tested For Virus Panel Testing According to 9 CFR 113.53
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.