missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Bovine Growth Serum (U.S.)

Product Code. 12300003
Change view
Click to view available options
Quantity:
500 mL
Processing:
Heat Inactivated
None
Unit Size:
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing Unit Size
12300003 500 mL Heat Inactivated 500mL
10453302 500 mL None 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12300003 Supplier Cytiva Supplier No. SH30541.03HI

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Currently Unavailable: We have no information when this product will be available.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Support cell culture growth with U.S.-sourced HyClone™ Bovine Growth Serum, a superior performing fetal bovine serum (FBS) replacement, consisting of bovine calf serum supplemented with chemically-defined components.

  • Filtered through sequential 100nm (0.1μm) pore-size rated filters
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Insulin, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Total Bilirubin, Sodium, Potassium, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, G
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Product Type Bovine Growth Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Heat Inactivated
Tested For Bluetongue, Bluetongue; Bovine; Adenovirus; Bovine Parvovirus; Bovine RSV; BVDV; Cytopathogenic Agents; Hemadsorbing Agents; Rabies Virus; Reovirus, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤ 10 mg/dL
Mycoplasma Detected Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.