missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cell Boost™ 1 Supplement
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- Chemically Defined (CD), Animal-Derived Component-Free (ADCF), and Protein-Free
- Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
- Supplements cell culture with amino acids, vitamins, and glucose
- Does not contain L-glutamine, poloxamer 188, or hydrolysates
- Manufactured to meet cGMP manufacturing standards and QC specifications
- Available as a ready-to-use liquid feed solution
Specifications
Specifications
| Cell Line | CHO, HEK293 |
| Form | Liquid |
| Product Type | Cell Boost Supplement |
| Format | Bottle |
| For Use With (Application) | Improving Productivity and Cell Growth in Fed-Batch Cultures |
| Product Line | HyClone |
| Quantity | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction