missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cell Boost™ 3 Supplement

Product Code. 16675795
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16675795 1000 mL 1 L
16524581 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16675795 Supplier Cytiva Supplier No. SH31115.02
  • ONLINE EXCLUSIVE PRICE
    185.00 €
    183.00 € / 1 L
    Save 2.00 €
    1% Off
 Restricted Item
Estimated Shipment: 16-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Fed-batch supplement with amino acids, vitamins, glucose, and trace elements for hybridoma and myeloma cells

  • Chemically Defined (CD), Animal-Derived Component-Free (ADCF), and Protein-Free
  • Supports fed-batch cultures with amino acids, vitamins, glucose, and trace elements
  • Developed for recombinant protein production with various cell lines including hybridoma and myeloma cell lines
  • Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
  • Manufactured to meet cGMP manufacturing standards and QC specifications
  • Available as a ready-to-use liquid feed solution
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hybridomas, Myelomas
Form Liquid
Product Type Cell Boost Supplement
Format Bottle
For Use With (Application) Improving Productivity and Cell Growth in Fed-Batch Cultures
Product Line HyClone
Quantity 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.