missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cell Boost™ 5 Supplement

Product Code. 16695795
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16695795 1000 mL 1 L
16544581 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16695795 Supplier Cytiva Supplier No. SH31117.02
  • ONLINE EXCLUSIVE PRICE
    280.00 €
    182.11 € / 1 L
    Save 97.89 €
    34% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fed-batch supplement with lipids, amino acids, vitamins, and growth factors for hybridoma, HEK 293, NS0, and CHO cells

  • Chemically Defined (CD), Animal-Derived Component-Free (ADCF), Protein-Free, and Hydrolysate-Free
  • Supports fed-batch cultures with lipids, amino acids, vitamins, and growth factors
  • Developed for recombinant protein production with various cell lines including Chinese Hamster Ovary (CHO) cell line, Human Embryonic Kidney (HEK) 293, NS0, and hybridoma cells
  • Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
  • Manufactured to meet cGMP manufacturing standards and QC specifications
  • Available as a ready-to-use liquid feed solution
TRUSTED_SUSTAINABILITY

Specifications

Cell Line CHO, HEK293, Hybridoma, NS0
Form Liquid
Product Type Cell Boost Supplement
Format Bottle
For Use With (Application) Improving Productivity and Cell Yield in Fed-Batch Cultures
Product Line HyClone
Quantity 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.