missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cell Boost™ 7a Supplement

Product Code. 16615805
Change view
Click to view available options
Quantity:
0.5 L
1 L
Form:
Liquid
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Form Unit Size
16615805 1 L Liquid 1 L
16564581 0.5 L Liquid 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16615805 Supplier Cytiva Supplier No. SH31119.02
  • ONLINE EXCLUSIVE PRICE
    348.00 €
    235.82 € / 1 L
    Save 112.18 €
    32% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A ready-to-use, powder media supplement optimized for high-yield recombinant protein production in CHO fed-batch processes

  • Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture
  • Supplements cell culture with amino acids, vitamins, salts, trace elements, and glucose
  • Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production
  • Designed to support modern manufacturing processes
  • Chemically defined (CD), animal-derived component-free (ADCF), and free of any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system
  • Formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines
  • Manufactured to meet cGMP manufacturing standards and QC specifications
TRUSTED_SUSTAINABILITY

Specifications

Description Without: L-glutamine, With: Poloxamer 188
Cell Line CHO
Form Liquid
Product Type Cell Culture Supplement
Sterility Non-sterile
For Use With (Application) High-Yield Recombinant Protein Production in CHO Fed-Batch Processes
Product Line HyClone
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.