missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cell Boost™ 7b Supplement
Description
- Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture
- A concentrated solution of amino acids with an alkaline pH
- Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production.
- Designed to support modern manufacturing processes
- Chemically defined (CD), animal-derived component-free (ADCF), and does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
- Intended for use in combination with HyClone ActiPro and ActiSM media as part of the ActiPro cell culture system
- Formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines
- Manufactured to meet cGMP manufacturing standards and QC specifications
Specifications
Specifications
| Content And Storage | Store 2°C to 8°C, Away From Light and Moisture |
| Description | Without: L-glutamine, poloxamer 188, and insulin |
| Form | Powder |
| Product Type | Amino Acid Supplement |
| Sterility | Non-sterile |
| For Use With (Application) | High-Yield Protein Production in CHO Fed-Batch Processes |
| Product Line | HyClone |
| Quantity | 500 mL |
Safety and Handling
Product Identifier
- Cell Boost 7b, without Poloxamer-188, without Insulin, without L-Glutamine
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: tyrosine
- UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?