missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone Characterized Fetal Bovine Serum (FBS), Australian Origin

Product Code. 11591841
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11591841 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11591841 Supplier Cytiva Supplier No. SH30084.03
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enhance growth of cells with Australia sourced HyClone Characterized Fetal Bovine Serum.

  • HyClone Characterized Fetal Bovine Serum (FBS), Australian Origin is sourced from Australian abattoirs approved by the U.S. Department of Agriculture (USDA) or Australian Quarantine and Inspection Service (AQIS).
  • The Australian methods of animal husbandry exhibit excellent animal nutrition and healthcare.
  • Australia is an island continent, thus making animal disease and control management easier than in most areas of the world.
  • Characterized FBS, Australian Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
  • Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles (true pool technology).
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Gamma Globulin Alkaline Phosphatase Lactate Dehydrogenase SGPT SGOT pH Albumin Blood Urea Nitrogen Creatinine Total Bilirubin Sodium Potassium Calcium Chloride Inorganic Phosphorus Iron TIBC Percent Saturation Glucose IgG
Certifications/Compliance USDA Regulations, AQIS
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
USDA Safety Testing Included USDA Inspected
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.