missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (FBS), U.S. Origin

Product Code. 16615735
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
HI, Irr - 25 to 40 doses
HI, Irr - 30 to 45 doses
Heat Inactivated, Irradiated
Irradiated
None
Unit Size:
1 L
100mL
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing Unit Size
16615735 100 mL Heat Inactivated, Irradiated 100mL
16261082 500 mL HI, Irr - 25 to 40 doses 500mL
16271082 500 mL HI, Irr - 30 to 45 doses 500mL
16625735 100 mL Irradiated 100mL
16635735 500 mL Irradiated 500mL
16645735 1000 mL None 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16615735 Supplier Cytiva Supplier No. SH30071.02IH

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Low in antibodies and high in growth factors to support robust growth and division of cells, including embryonic stem cells.

  • Additional traceability testing of HyClone™ Fetal Bovine Serum is performed by a collaborator, Oritain
  • Low in antibodies and high in growth factors
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Each lot is subjected to virus panel testing according to 9 CFR 113.53
  • Characterized FBS is assayed for many components, including gamma globulin, glucose, and IgG
  • Supports robust growth and division of cells, including embryonic stem cells
  • Produced using true pool technology to minimize bottle-to-bottle variability
  • Complete traceability is provided back to the original source, as certified by the International Serum Industry Association (ISIA)
  • Sourced in the USA
TRUSTED_SUSTAINABILITY

Specifications

Country of Origin United States
Form Serum
Product Type Fetal Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Grade Characterized
Processing Heat Inactivated, Irradiated
Tested For Virus Panel Testing According to 9 CFR 113.53
Volume (Metric) 100 mL
Product Line HyClone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.