missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S.)

Product Code. 16281082
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
HI, Irr - 25 to 40 doses
Heat Inactivated
Irradiated
Irradiated (25 to 40 doses)
Irradiated (30 to 45 doses)
None
Unit Size:
100mL
500mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing Unit Size
16281082 500 mL Irradiated (25 to 40 doses) 500mL
11521831 100 mL None 100mL
17296886 100 mL HI, Irr - 25 to 40 doses 100mL
17206896 100 mL Irradiated (25 to 40 doses) 100mL
11531831 500 mL None 500mL
12379802 500 mL Heat Inactivated 500mL
11306060 500 mL Irradiated 500mL
16291082 500 mL Irradiated (30 to 45 doses) 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 16281082 Supplier Cytiva Supplier No. SH30071.03IR2540
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Used as serum-supplement for the in vitro cell culture of eukaryotic cells

  • Enhance growth of cells
  • Tested for endotoxin, hemoglobin, and additional biochemical assays
  • Filtered through serial 100 nm pore-size rated filters
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Country of Origin United States
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Grade Characterized
Processing Irradiated (25 to 40 doses)
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 25 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 25 mg/dL
Product Line HyClone
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.