missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cosmic Calf Serum (New Zealand origin)

A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.

Brand:  Cytiva SH30413.03IR

Code : 85

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12624227

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

All New Zealand calf serum products are carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases. We follow the same standards in processing of our calf serum as with our fetal bovine serum (FBS) to enable offering the highest quality and traceability.Cosmic Calf Serum, New Zealand Origin is filtered through sequential 100 nm (0.1 µm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.

Specifications

Specifications

Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
1 L
Sequential-filtered, 100 nm
≤ 25 EU/mL
≤ 25 mg/dL
USDA Inspected
USDA Regulations
FBS Replacement Serum
Sterile
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
No
Yes
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen
Sonderangebote

Sonderangebote