missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)

Product Code. 10287963
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Gamma Irradiated
Heat Inactivated
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
None
Unit Size:
1000mL
100mL
500mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing Unit Size
10287963 100 mL None 100mL
11556541 1000 mL Gamma Irradiated 1000mL
10136253 500 mL None 500mL
10176363 1000 mL None 1000mL
12389842 500 mL Heat Inactivated 500mL
16231102 500 mL Irradiated, 25 to 40 doses 500mL
16241102 1000 mL Irradiated, 25 to 40 doses 1000mL
16251102 1000 mL Irradiated, 30 to 45 doses 1000mL
12369842 500 mL Gamma Irradiated 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10287963 Supplier Cytiva Supplier No. SH30087.02

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Currently Unavailable: We have no information when this product will be available.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains high quality iron-supplemented bovine calf serum with added growth promoting factors.

  • Age at time of collection: typically 16 - 22 weeks
  • Iron-supplemented with additional growth promoting components
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Insulin, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Phosphorus, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description Calf Serum; Cosmic Calf; 100 mL
Additive Iron and naturally derived components
Product Type Calf Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm, Gamma-irradiated
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.