missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Gamma Irradiated
Heat Inactivated
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
None
Unit Size:
1000mL
100mL
500mL
Description
- Age at time of collection: typically 16 - 22 weeks
- Iron-supplemented with additional growth promoting components
Specifications
Specifications
| Additional Assays | Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Insulin, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Phosphorus, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein |
| Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
| Description | Gamma Irradiated Calf Serum |
| Additive | Iron and naturally derived components |
| Product Type | Calf Bovine Serum |
| Quantity | 1000 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm, Gamma-irradiated |
| Processing | Gamma Irradiated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction