missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Powder
Shop All Thermo Scientific Chemicals ProductsDescription
Buffer cell culture with HyClone™ Dulbecco's Phosphate Buffered Saline (PBS), Powder.
Specifications
Specifications
| Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
| Content And Storage | 2° to 8° C |
| Form | Powder |
| Sterility | Non-sterile |
| Without Additives | No Calcium, No Magnesium |
| Quantity | 10 L |
Safety and Handling
Product Identifier
- Sodium sulfide nonahydrate
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H290-May be corrosive to metals.
- EUH031-Contact with acids liberates toxic gas.
- EUH071-Corrosive to the respiratory tract.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?