missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution

Product Code. 10317052
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
1000 mL
6 x 500 mL
10 L
20 L
6 x 1000 mL
Concentration:
1 X
10 X
With Additives:
Calcium, Magnesium
None
Without Additives:
No Calcium, No Magnesium
No Calcium, No Magnesium, No Phenol Red
No Phenol Red
Unit Size:
10 L
1000mL
100mL
20 L
500mL
Pack of 6
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Concentration With Additives Without Additives Unit Size
10317052 500 mL 1 X None No Calcium, No Magnesium, No Phenol Red 500mL
11581811 100 mL 1 X None No Calcium, No Magnesium, No Phenol Red 100mL
10687974 6 x 1000 mL 1 X None No Calcium, No Magnesium, No Phenol Red Pack of 6
10540853 1000 mL 1 X Calcium, Magnesium No Phenol Red 1000mL
11591811 10 L 1 X None No Calcium, No Magnesium 10 L
11511821 6 x 500 mL 1 X None No Calcium, No Magnesium, No Phenol Red Pack of 6
10044823 6 x 500 mL 1 X Calcium, Magnesium No Phenol Red Pack of 6
10708144 1000 mL 1 X None No Calcium, No Magnesium, No Phenol Red 1000mL
10798134 500 mL 1 X Calcium, Magnesium No Phenol Red 500mL
12882946 1 L 10 X None No Calcium, No Magnesium, No Phenol Red 1000mL
10021352 500 mL 10 X None No Calcium, No Magnesium, No Phenol Red 500mL
11586511 20 L 1 X None No Calcium, No Magnesium 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 10317052 Supplier Cytiva Supplier No. SH30028.02
  • ONLINE EXCLUSIVE PRICE
    27.46 €
    26.00 € / 500mL
    Save 1.46 €
    5% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Buffer cell culture with PBS in solution.

  • Format: Liquid
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Manufactured under cGMP and ISO 9001 certified facilities
Content And Storage 15° to 30°C
Description HyClone Dulbecco's Phosphate Buffered Salines (DPBS) Without Calcium, Magnesium, Phenol Red
Concentration 1 X
Form Liquid
Product Type Dulbecco's Phosphate Buffered Saline
Sterility Sterile
Sterilization Method 0.1 μm Sterile-filtered
With Additives None
Without Additives No Calcium, No Magnesium, No Phenol Red
Endotoxin Level ≤0.025EU/mL
For Use With (Application) Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
Osmolality 276±10mOsm/kg H2O
pH 7 to 7.6
Quantity 500 mL
Regulatory Status RUO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.