missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Bovine Serum (U.S.), Charcoal/Dextran Treated

Product Code. 10611235
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Processing:
Gamma Irr, 25 to 40 doses
Gamma Irradiated
Heat Inactivated
None
Heat Inactivated:
No
Yes
Unit Size:
100mL
500mL
50mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing Heat Inactivated unitSize
10611235 500 mL None No 500mL
12339782 500 mL Gamma Irradiated No 500mL
12329782 500 mL Heat Inactivated Yes 500mL
12319782 100 mL Heat Inactivated Yes 100mL
12634217 50 mL Heat Inactivated Yes 50mL
12389772 50 mL None No 50mL
11571821 100 mL None No 100mL
16201082 500 mL Gamma Irr, 25 to 40 doses No 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10611235 Supplier Cytiva Supplier No. SH30068.03
  • ONLINE EXCLUSIVE PRICE
    770.00 €
    762.00 € / 500mL
    Save 8.00 €
    1% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Augment growth of cells with U.S.-sourced HyClone™ Charcoal/Dextran Treated Fetal Bovine Serum (FBS), containing reduced levels of many hormones and growth factors.

  • Processed using a proprietary charcoal/dextran treatment that has been shown to reduce the levels of many hormones and growth factors
  • This serum is assayed for a variety of components before and after treatment to ensure the efficacy of the treatment
  • Charcoal/dextran treatment has been shown to be effective in reducing steroid levels for use of the serum in receptor studies or estrogen-related investigations
  • Other studies indicate that charcoal/dextran treatments may be used to minimize lot-to-lot serum variability
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays 1, 25 Dihydroxy Vitamin D, 17 Beta Estradiol, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcitonin, Calcium, Carotene, Chloride, Corticosterone, Cortisol, Creatinine, Estrone, Folate, FSH, Glucose, IgG, Inorganic Phosphorus, Insulin, Lactate Dehydrogenase, LH, Niacin, Potassium, Progesterone, Retinol, SGOT, SGPT, Sodium, T3, T4, Testosterone, Tocopherol, Total Bilirubin, Total Protein, TSH, Vitamin C
Certifications/Compliance EMEA Certified
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description Charcoal/Dextran Treated Fetal Bovine Serum, US Origin, None
Country of Origin United States
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Grade Specialty filtered
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 20 mg/dL
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.