Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Charcoal/Dextran Treated
Augment growth of cells with U.S.-sourced HyClone™ Charcoal/Dextran Treated Fetal Bovine Serum (FBS), containing reduced levels of many hormones and growth factors.
Brand: Cytiva SH30068.03IR25-40
516.74 EUR valid until 2024-12-31
Use promo code "21878" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
- Processed using a proprietary charcoal/dextran treatment that has been shown to reduce the levels of many hormones and growth factors
- This serum is assayed for a variety of components before and after treatment to ensure the efficacy of the treatment
- Charcoal/dextran treatment has been shown to be effective in reducing steroid levels for use of the serum in receptor studies or estrogen-related investigations
- Other studies indicate that charcoal/dextran treatments may be used to minimize lot-to-lot serum variability
- Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
- Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
Specifications
25 Dihydroxy Vitamin D, 17 Beta Estradiol, Albumin, Alkaline Phosphatase | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Charcoal/Dextran filtered | |
Sterile | |
Specialty filtered | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV | |
No | |
No |
EMEA Certified | |
United States | |
500 mL | |
Sequential-filtered, 100 nm | |
Gamma Irradiated, 25 to 40 doses | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
USDA Inspected |