missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Bovine Serum (U.S.), Standard

Product Code. 11511851
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Gamma Irradiated
Heat Inactivated
Irradiated, 25 to 40 doses
None
Unit Size:
100mL
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing missing translation for 'unitSize'
11511851 100 mL None 100mL
12399842 100 mL Heat Inactivated 100mL
16271102 500 mL Irradiated, 25 to 40 doses 500mL
11521851 500 mL None 500mL
12309852 500 mL Heat Inactivated 500mL
12339852 500 mL Gamma Irradiated 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11511851 missing translation for 'mfr' Cytiva missing translation for 'supplierNo' SH30088.02
  • ONLINE EXCLUSIVE PRICE
    162.00 €
    160.00 € / 100mL
    Save 2.00 €
    1% Off
View all promotions
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Supplement cell culture growth with U.S. sourced quality cell culture-grade Cytiva HyClone™ Standard Fetal Bovine Serum (FBS)

  • Support cell culture growth
  • Standard grade
  • Aseptically packaged in Class 100 cleanroom
TRUSTED_SUSTAINABILITY

Spezifikation

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description Standard Fetal Bovine Serum, US Origin
Country of Origin United States
Product Type Fetal Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Grade Standard
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration For Information Only
Heat Inactivated No
Hemoglobin Concentration For Information Only
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.