missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ FetalClone™ II Serum (U.S.)

Product Code. 16291072
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
HI, Irr - 25 to 40 doses
Heat Inactivated
Irradiated
Irradiated (25 to 40 doses)
Irradiated (30 to 45 doses)
Unit Size:
100mL
500mL
Product Code. Quantity Processing unitSize
16291072 500 mL Irradiated (25 to 40 doses) 500mL
12339772 100 mL Heat Inactivated 100mL
17662065 500 mL HI, Irr - 25 to 40 doses 500mL
14850841 500 mL Irradiated 500mL
17256886 500 mL Irradiated (30 to 45 doses) 500mL
5 options
This item is not returnable. View return policy

Product Code. 16291072

Brand: Cytiva SH30066.03IR2540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimized for the growth of CHO cells and derivatives.

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
  • demonstrated performance with a variety of cell lines, including hybridomas, CHO, BHK-21, NS0, MRC-5, and Vero cells
  • TRUSTED_SUSTAINABILITY

    Specifications

    Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
    Country of Origin United States
    Product Type FBS Replacement Serum
    Quantity 500 mL
    Sterility Sterile
    Sterilization Method Sequential-filtered, 100 nm
    Processing Irradiated (25 to 40 doses)
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 10 EU/mL
    Heat Inactivated No
    Hemoglobin Concentration ≤ 20 mg/dL
    IgG levels Typically less than 2 μg/mL
    Product Line HyClone
    Mycoplasma Detected No
    USDA Safety Testing Included USDA Inspected
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.