missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ FetalClone™ II Serum (U.S.)

Product Code. 17662065
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
HI, Irr - 25 to 40 doses
Heat Inactivated
Irradiated
Irradiated (25 to 40 doses)
Irradiated (30 to 45 doses)
Unit Size:
100mL
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing Unit Size
17662065 500 mL HI, Irr - 25 to 40 doses 500mL
12339772 100 mL Heat Inactivated 100mL
14850841 500 mL Irradiated 500mL
16291072 500 mL Irradiated (25 to 40 doses) 500mL
17256886 500 mL Irradiated (30 to 45 doses) 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17662065 Supplier Cytiva Supplier No. SH30066.03IH2540
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Optimized for the growth of CHO cells and derivatives.

  • Economical alternative to FBS
  • Performance comparable with FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
  • demonstrated performance with a variety of cell lines, including hybridomas, CHO, BHK-21, NS0, MRC-5, and Vero cells
  • TRUSTED_SUSTAINABILITY

    Specifications

    Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
    Country of Origin United States
    Product Type FBS Replacement Serum
    Quantity 500 mL
    Sterility Sterile
    Sterilization Method Sequential-filtered, 100 nm
    Processing HI, Irr - 25 to 40 doses
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 10 EU/mL
    Heat Inactivated Yes
    Hemoglobin Concentration ≤ 20 mg/dL
    IgG levels Typically less than 2 μg/mL
    Product Line HyClone
    Mycoplasma Detected No
    USDA Safety Testing Included USDA Inspected
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.