missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ FetalClone™ III Serum (U.S.)
Designed for fibroblast applications, supplement cell culture growth with U.S.-sourced HyClone™ FetalClone™ III Serum, the most widely applicable fetal bovine serum (FBS) alternative.
Brand: Cytiva SH30109.02HI
Code : 84
Additional Details : Weight : 0.10000kg
Description
- Economical alternative to FBS
- Performance comparable with FBS
- High lot-to-lot consistency
- Low endotoxin and hemoglobin levels
Specifications
Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
FBS Replacement Serum | |
Sterile | |
Heat Inactivated | |
≤ 10 EU/mL | |
≤ 20 mg/dL | |
HyClone™ | |
USDA Inspected |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
United States | |
100 mL | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Yes | |
Typically less than 2 μg/mL | |
No |
Sécurité et traitement
Mention de danger
Informations supplémentaires
- EUH210-Safety data sheet available on request.
Informations supplémentaires
- UNKNOWN ACUTE-9.9 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 9.9% of components with unknown hazards to the aquatic environment.