missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ HyPure Water For Injection (WFI) Quality Water, Cytiva

Product Code. 12379872
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
12379872 100 mL 100mL
1 options
This item is not returnable. View return policy

Product Code. 12379872

Brand: HyClone products Cytiva SH30221.18

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Supports Cell Culture Growth and Biomanufacturing Processes

  • HyPure WFI Quality Water undergoes extensive purification to meet or exceed the stringent specifications of the United States Pharmacopeia (USP)
  • Actual test results on a CoA are not taken from system samples, but must be from actual packaged samples of the given lot
  • Manufactured in ISO 9001 certified facility
  • Lot-specific certificate of analysis (CoA) for each lot of water produced
  • Intended for further manufacturing or research use
  • Not for diagnostic or therapeutic use
TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5
Melting Point 0°C
Boiling Point 100°C
Description Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Molecular Formula H2O
MDL Number MFCD00011332
Merck Index 1310098
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Sterility Sterile
Density 1 g/cm3
Certificate of Analysis Included
Quantity 100 mL
Physical Form Liquid
Product Line HyClone™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.