missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HyClone™ HyQTase Cell Detachment Reagent
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Descripción
- Ultra-filtered, non-mammalian alternative to trypsin that enables gentle, rapid cell detachment of a broad array of adherent cells and tissues for primary cell culture
- Naturally derived complex of proteolytic and collagenolytic enzymes
- Ideal for use in both serum-free and serum-containing cell culture systems with a broad array of cell lines
Especificaciones
Especificaciones
| Content And Storage | Store frozen; stable for up to 60 days at 4°C. |
| Description | HyQTase Cell Detachment Solution, in DPBS with EDTA (non-mammalian) |
| With Additives | EDTA |
| Quantity | 100 mL |
| Source | Non-mammalian |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido