missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum (New Zealand)

Product Code. p-7087599
Click to view available options
Quantity:
1000 mL
Processing:
Irradiated
Irradiated (25 to 40 doses)
None
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 16605765

Brand: Cytiva SH30414.03

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A cost-effective alternative to Fetal Bovine Serum (FBS)

  • Sourced from New Zealand with complete traceability back to original source
  • Age of animal at time of collection less than 10 days old
  • Filtered through sequential 100 nm pore size-rated filters
  • Virus panel testing according to 9 CFR 113.53
  • Low in antibodies and high in growth factors
  • Supplemented with iron
  • Each serum lot is pooled using true pool technology to ensure uniformity and consistency between bottles
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, and IgG
Content And Storage Store at or below -10°C
Country of Origin New Zealand
Form Liquid
Product Type Calf Bovine Serum
Quantity 1000 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 25 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 25 mg/dL
Iron Supplemented Yes
Product Line HyClone
USDA Safety Testing Included Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.