missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum, US Origin

Product Code. 16695735
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16695735 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16695735 Supplier Cytiva Supplier No. SH30072.04IRBI35

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Contains additional iron that boosts serum transferrin levels to physiological levels

  • Supplemented with iron and growth-promoting factors
  • Age of animal at time of collection approximately 16-24 weeks
  • Virus panel testing according to 9 CFR 113.53
  • Low in antibodies and high in growth factors
  • Sourced in the USA with complete traceability back to original source
  • A cost-effective alternative to FBS
TRUSTED_SUSTAINABILITY

Especificaciones

Certifications/Compliance Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Country of Origin United States
Quantity 1000 mL
Processing Irradiated
Endotoxin Concentration <10 EU/mL
For Use With (Application) Supplementing Cell Culture Growth
Iron Supplemented Yes
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.