missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum, US Origin

Product Code. 16211092
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
HI, Irr - 25 to 40 doses
Irradiated
Irradiated, Heat Inactivated
Non-irradiated
None
Unit Size:
1 L
100mL
500mL
Product Code. Quantity Processing unitSize
16211092 1000 mL HI, Irr - 25 to 40 doses 1 L
12829447 500 mL Irradiated, Heat Inactivated 500mL
16685735 1000 mL Irradiated 1 L
16605745 1000 mL Non-irradiated 1 L
16625755 100 mL None 100mL
16635755 1000 mL None 1 L
6 options
This item is not returnable. View return policy

Product Code. 16211092

Brand: Cytiva SH30072.04IH2540

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contains additional iron that boosts serum transferrin levels to physiological levels

  • Supplemented with iron and growth-promoting factors
  • Age of animal at time of collection approximately 16-24 weeks
  • Virus panel testing according to 9 CFR 113.53
  • Low in antibodies and high in growth factors
  • Sourced in the USA with complete traceability back to original source
  • A cost-effective alternative to FBS
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Age 16 to 22 weeks
Country of Origin United States
Product Type Calf Bovine Serum
Quantity 1000 mL
Sterilization Method Sequential-filtered, 100 nm
Source Formula-Fed Veal Calves
Processing HI, Irr - 25 to 40 doses
Tested For Virus Panel Testing According to 9 CFR 113.53
Endotoxin Concentration <10 EU/mL
For Use With (Application) Supplementing Cell Culture Growth
Heat Inactivated Yes
Hemoglobin Concentration <10 mg/dL
Iron Supplemented Yes
Product Line HyClone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.