missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iron-Supplemented Calf Serum, U.S. origin

Product Code. 16201092
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Gamma Irradiated
Heat Inactivated
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
None
Unit Size:
1 L
1000mL
100mL
500mL
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing unitSize
16201092 500 mL Irradiated, 25 to 40 doses 500mL
12349812 500 mL Gamma Irradiated 500mL
16221092 1000 mL Irradiated, 25 to 40 doses 1 L
10761454 100 mL None 100mL
16231092 1000 mL Irradiated, 30 to 45 doses 1000mL
12319812 100 mL Heat Inactivated 100mL
15715533 1000 mL Heat Inactivated 1000mL
11551831 500 mL None 500mL
12339812 500 mL Heat Inactivated 500mL
12839447 1000 mL Gamma Irradiated 1 L
10339953 1000 mL None 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 16201092 Supplier Cytiva Supplier No. SH30072.03IR2540
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

A cost-effective alternative to Fetal Bovine Serum (FBS). Contains additional iron to boost serum transferrin to physiological levels.

HyClone Iron-Supplemented Bovine Calf Serum, US Origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. This serum is carefully supplemented with iron to load serum transferrin levels to physiological levels. After supplementation, the serum contains three to four times as much available iron and transferrin as fetal bovine serum (FBS) or equine serum.

TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Irradiated, 25 to 40 doses
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Heat Inactivated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.