missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum (New Zealand)

Product Code. 10761274
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Processing:
Heat Inactivated
Irradiated
Irradiated (25 to 40 doses)
None
Unit Size:
1 L
1000mL
500mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing unitSize
10761274 500 mL None 500mL
12330043 500 mL Irradiated 500mL
11582830 1000 mL None 1000mL
12694217 1000 mL Heat Inactivated 1000mL
12684217 1000 mL Irradiated 1000mL
17236946 1000 mL Irradiated (25 to 40 doses) 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10761274 Supplier Cytiva Supplier No. SH30626.02

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A cost-effective alternative to Fetal Bovine Serum (FBS).

  • Ensured safety against bovine diseases
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Ensured uniformity and consistency between bottles
  • TRUSTED_SUSTAINABILITY

    Specifications

    Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
    Country of Origin New Zealand
    Product Type Newborn Calf Bovine Serum
    Quantity 500 mL
    Sterility Sterile
    Sterilization Method Sequential-filtered, 100 nm
    Processing None
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 25 EU/mL
    Heat Inactivated No
    Hemoglobin Concentration ≤ 25 mg/dL
    Product Line HyClone
    USDA Safety Testing Included Yes
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.