missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Irradiated Calf Serum, U.S. Origin

Product Code. 16615745
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16615745 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16615745 Supplier Cytiva Supplier No. SH30073.02IR

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin

  • Age of animal at time of collection ranges from 16 to 24 weeks
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
TRUSTED_SUSTAINABILITY

Specifications

Age 16 to 24 weeks
Country of Origin United States
Product Type Calf Bovine Serum
Quantity 100 mL
Sterilization Method Sequential-filtered, 100 nm
Processing Irradiated
Tested For Virus Panel Testing According to 9 CFR 113.53
Heat Inactivated No
Product Line HyClone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.