missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ L-Glutamine, Powder
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Description
- Support the growth of cells that have high energy demands
- Synthesize large amounts of proteins and nucleic acids
Specifications
Specifications
| Amino Acids | L-Glutamine |
| Form | Powder |
| Product Type | L-Glutamine |
| For Use With (Application) | Supplementing Serum- and Protein-Free Media |
| Product Line | HyClone |
| Quantity | 25 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction