missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Porcine Serum (New Zealand)

Product Code. 17236916
Change view
Click to view available options
Quantity:
1 L
1 L (QC Samples)
Processing:
Irradiated
Non-irradiated
Unit Size:
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing unitSize
17236916 1 L (QC Samples) Irradiated 1 L
16695785 1 L Non-irradiated 1 L
16685785 1 L Irradiated 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17236916 Supplier Cytiva Supplier No. SH30908.04SAM

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used to enhance cell culture performance, cost-efficient alternatives to Fetal Bovine Serum (FBS)

Cytiva's porcine serum is sourced from New Zealand, filtered in the U.S. through multiple 200 nm pore size-rated filters and is gamma irradiated at 25 to 40 kGy after filtration.
TRUSTED_SUSTAINABILITY

Specifications

Country of Origin New Zealand
Form Liquid
Product Type Porcine Serum
Quantity 1 L (QC Samples)
Sterilization Method Sequential Triple-filtered, 200 nm
Processing Irradiated
Endotoxin Concentration ≤ 100 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 50 mg/dL
Product Line HyClone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.