missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin

Product Code. 16211112
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16211112 500 mL 500mL
16241112 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16211112 Supplier Cytiva Supplier No. SH30401.01IR3040
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

An excellent and cost-effective alternative to Fetal Bovine Serum (FBS)

  • Supplements cell culture growth
  • Sourced from animals that are typically less than 10 days old
  • Manufactured in accordance with cGMP guidelines at an ISO 9001: 2008 accredited facility
  • Conforms to USDA regulations
  • Traceable
  • Filtered
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in Freezer, <20°C
Age <10 days
Country of Origin New Zealand
Form Serum
Product Type Newborn Calf Bovine Serum
Quantity 500 mL
Sterilization Method Sequential-filtered, 100 nm
Processing Irradiated (30 to 40 kGy)
Volume (Metric) 500 mL
For Use With (Application) Supplementing Cell Culture Growth
Product Line HyClone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.