missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin

A cost-effective alternative to Fetal Bovine Serum (FBS).

Brand:  Cytiva SH30401.01IR25-40

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 16201112

  • 91.70 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available
View alternative products

Description

Description

  • Carefully collected, processed, and filtered in New Zealand
  • Ensured safety against bovine diseases
  • We follow the same standards in processing of our calf serum as with our fetal bovine serum (FBS) to enable offering the highest quality and traceability
  • HyClone Newborn Calf Serum, New Zealand Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
  • Specifications

    Specifications

    Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG
    Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
    Sterile
    Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    No
    Yes
    USDA Regulations
    500 mL
    Irradiated, 25 to 40 doses
    ≤ 25 EU/mL
    Yes
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers