missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Calf Serum (U.S.)

Product Code. 11571851
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Irradiated, 25 to 40 doses
None
Unit Size:
1 L
1000mL
100mL
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing unitSize
11571851 100 mL None 100mL
11556551 1000 mL None 1000mL
11581851 500 mL None 500mL
16291102 500 mL Irradiated, 25 to 40 doses 500mL
16605755 1000 mL Irradiated, 25 to 40 doses 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11571851 Supplier Cytiva Supplier No. SH30118.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.

  • Filtered through sequential 100nm filters and sourced from animals that are typically less than 10 days old
TRUSTED_SUSTAINABILITY

Specifications

Description Newborn Calf Serum (U.S.)
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Processing None
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.