missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media

Product Code. 10721694
Change view
Click to view available options
Quantity:
1000 mL
5 L
10 L
6 x 1000 mL
With Additives:
Glutamine, Poloxamer 188, Sodium Bicarbonate
None
Without Additives:
No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate
No Phenol Red, No HEPES
Product Type:
PF CHO LS
PF-CHO LS
PF-CHO MPS
Unit Size:
10 L
1000mL
5 L
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity With Additives Without Additives Product Type Unit Size
10721694 5 L None No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate PF-CHO MPS 5 L
10494085 10 L None No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate PF-CHO MPS 10 L
11571911 6 x 1000 mL Glutamine, Poloxamer 188, Sodium Bicarbonate No Phenol Red, No HEPES PF CHO LS Pack of 6
10443295 1000 mL Glutamine, Poloxamer 188, Sodium Bicarbonate No Phenol Red, No HEPES PF-CHO LS 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10721694 Supplier Cytiva Supplier No. SH30333.01
  • ONLINE EXCLUSIVE PRICE
    173.00 €
    126.00 € / 5 L
    Save 47.00 €
    27% Off
 Restricted Item
Estimated Shipment: 05-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.

  • Designed for high cell yield and recombinant protein production
  • Allow for direct or sequential adaptation
  • Support growth in multiple cell culture systems
  • Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
  • TRUSTED_SUSTAINABILITY

    Specifications

    Form Powder
    Product Type PF-CHO MPS
    Sterility Non-sterile
    With Additives None
    Without Additives No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate
    For Use With (Application) Gowth of multiple Chinese hamster ovary (CHO) cell clones and production of a variety of recombinant proteins with minimal adaptation
    Quantity 5 L
    Certifications/Compliance cGMP
    Content And Storage 2° to 8°C, away from light
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.