missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SFM4CHO™ Media

Product Code. 10483522
Change view
Click to view available options
Quantity:
500 mL
1000 mL
5 L
With Additives:
Glutamine, Sodium Bicarbonate, Poloxamer 188
Sodium Bicarbonate, Poloxamer 188
Without Additives:
No Glutamine, No Phenol Red, No HEPES
No Glutamine, No Sodium Bicarbonate, No Poloxamer 188, No Phenol Red, No HEPES
No Phenol Red, No HEPES
Form:
Liquid
Powder
Unit Size:
1000mL
5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity With Additives Without Additives Form Unit Size
10483522 1000 mL Glutamine, Sodium Bicarbonate, Poloxamer 188 No Phenol Red, No HEPES Liquid 1000mL
10580083 5 L - No Glutamine, No Sodium Bicarbonate, No Poloxamer 188, No Phenol Red, No HEPES Powder 5 L
10631243 500 mL Glutamine, Sodium Bicarbonate, Poloxamer 188 No Phenol Red, No HEPES Liquid 500mL
10376852 1000 mL Sodium Bicarbonate, Poloxamer 188 No Glutamine, No Phenol Red, No HEPES Liquid 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10483522 Supplier Cytiva Supplier No. SH30549.02
  • ONLINE EXCLUSIVE PRICE
    90.60 €
    57.51 € / 1000mL
    Save 33.09 €
    36% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for high performance in a variety of culture vessels, including bioreactors. HyClone™ SFM4CHO™ Media is a protein-free formulation that contains no components of bovine origin.

  • Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type SFM4CHO Liquid Media
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Sodium Bicarbonate, Poloxamer 188
Without Additives No Phenol Red, No HEPES
Quantity 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Description SFM4CHO media
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.