missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SFM4HEK293™ Media

Product Code. 10500283
Change view
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1000mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10500283 1000 mL 1000mL
10061502 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10500283 Supplier Cytiva Supplier No. SH30521.02
  • ONLINE EXCLUSIVE PRICE
    149.00 €
    63.00 € / 1000mL
    Save 86.00 €
    57% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Support production of adenoviral vectors and recombinant proteins in HEK 239 cultures with HyClone™ SFM4HEK293™ Media, a versatile, protein-free cell culture media containing no animal-derived components.

TRUSTED_SUSTAINABILITY

Specifications

Classification None
Form Liquid
Product Type HEK293 Medium
Sterility Sterile
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Poloxamer 188, Sodium Bicarbonate
Without Additives No Protein, No Phenol Red, No HEPES
Quantity 1000 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Description SFM4HEK293 Media
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.