missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SFM4Transfx-293 Media With Poloxamer 188 and Sodium Bicarbonate

Product Code. 11532910
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11532910 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11532910 Supplier Cytiva Supplier No. SH30860.04

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Promotes transfection using lipofection, polymer, or similar methods in HEK 293 cell culture

  • Developed to support high transfection efficiency, cell density and productivity in suspension cultures
  • Allows for direct or sequential adaptation
  • Serum-free, animal-derived, component-free
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • For high cell yield and recombinant protein/vector production
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C away from light
Description Contains poloxamer 188 and sodium bicarbonate, Without L-glutamine, phenol red and HEPES
For Use With (Application) For promoting high transfection efficiency, cell density and productivity in suspension cultures
Packaging Type Bag
Product Type HyClone SFM4Transfx-293 Media
Quantity 5 L
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.