missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Standard Fetal Bovine Serum, U.S. Origin

Product Code. 16261102
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16261102 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16261102 Supplier Cytiva Supplier No. SH30088.03IH2540
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Enhances cell culture growth in research applications, an economical alternative to characterized and defined Fetal Bovine Serum

  • Produced using true pool technology for uniformity and consistency from bottle to bottle
  • Filtered through three sequential 100 nm pore size-rated filters
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
  • Sourced in the USA with complete traceability back to original source
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, and IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Content And Storage Store at or below -10°C
Country of Origin United States
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterilization Method Sequential-filtered, 100 nm
Grade Standard
Processing Irradiated, Heat Inactivated
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, and Reovirus
Volume (Metric) 500 mL
Heat Inactivated Yes
Product Line HyClone
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.