missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ US Bovine Calf Serum

Product Code. 12849447
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. Quantity unitSize
12849447 500 mL 500mL
1 options
This item is not returnable. View return policy

Product Code. 12849447

Brand: Cytiva SH30073.03IH

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A cost effective alternative to Fetal Bovine Serum (FBS). Cytiva HyClone™ US Bovine Calf Serum is defined, irradiated, heat inactivated serum of US origin that is filtered through three sequential 0.1μm pore size-rated filters.

HyClone Bovine Calf Serum, U.S. origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.

  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
  • Age of animal at time of collection approximately 16-24 weeks
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Insulin Gamma Globulin Alkaline Phosphatase Lactate Dehydrogenase SGPT SGOT pH Albumin Blood Urea Nitrogen Creatinine Total Bilirubin Sodium Potassium Calcium Chloride Inorganic Phosphorus Iron TIBC Percent Saturation Glucose IgG
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description Heat Inactivated; 500 mL
Species Bovine
Age 16 to 24 weeks
Country of Origin United States
Product Type Calf Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Tested For Bluetongue Bovine Adenovirus Bovine Parvovirus Bovine RSV BVDV Cytopathogenic Agents Hemadsorbing Agents Rabies Virus Reovirus
Heat Inactivated Yes
USDA Safety Testing Included Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.