missing translation for 'onlineSavingsMsg'
Learn More

Hydantoin, 99%

Product Code. 11494104
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11494104

Brand: Thermo Scientific Alfa Aesar A12486.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 461-72-3 | C3H4N2O2 | 100.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-72-3
Molecular Formula C3H4N2O2
Molecular Weight (g/mol) 100.08
MDL Number MFCD00005259
InChI Key WJRBRSLFGCUECM-UHFFFAOYSA-N
Synonym hydantoin, 2,4-imidazolidinedione, glycolylurea, imidazole-2,4 3h,5h-dione, dantochlor, 2,4 3h,5h-imidazoledione, hydantoine, ccris 6532, 2-imidazolin-4 or 5-one, 2-hydroxy, 4h-imidazole-2,5-diol
PubChem CID 10006
ChEBI CHEBI:27612
IUPAC Name imidazolidine-2,4-dione
SMILES O=C1CNC(=O)N1

Specifications

Melting Point 221°C to 223°C
Quantity 100 g
Beilstein 110598
Merck Index 14,4761
Formula Weight 100.08
Percent Purity 99%
Chemical Name or Material Hydantoin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.