missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine-1,2-dicarboxamide, TRC

Product Code. 30393644
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393644 1 g -
30390065 5 g -
30393564 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393644 Supplier Toronto Research Chemicals Supplier No. TRCH0547101G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-21-4
Molecular Formula C2H6N4O2
Molecular Weight (g/mol) 118.1
Synonym Trazodone Impurity 21 (Biurea)
IUPAC Name (carbamoylamino)urea
SMILES NC(=O)NNC(=O)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H6N4O2/c3-1(7)5-6-2(4)8/h(H3,3,5,7)(H3,4,6,8)
Formula Weight 118.0491
Chemical Name or Material Hydrazine-1,2-dicarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.