missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine hydrate, 55% (Hydrazine, 35%)

Product Code. 10348830 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10348830

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10217-52-4 | H4N2 | 32.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10217-52-4, 7732-18-5
Molecular Formula H4N2
Molecular Weight (g/mol) 32.05
MDL Number MFCD00149931
InChI Key OAKJQQAXSVQMHS-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
SMILES NN

Specifications

Boiling Point 109.4°C
Melting Point -65.0°C
Physical Form Liquid
Quantity 500 mL
Assay Percent Range 55%
Linear Formula H2NNH2·xH2O
Fieser 01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174
Flash Point >100°C
Merck Index 14,4771
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,insoluble in chloroform and ether
IUPAC Name hydrazine;hydrate
Formula Weight 32.04
Percent Purity 55% (Hydrazine,35%)
Packaging Glass bottle
Refractive Index 1.3870 to 1.3910
Specific Gravity 1.023
Density 1.0230g/mL
Chemical Name or Material Hydrazine hydrate, 55% (Hydrazine 35%)
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Identificativo prodotto
  • Hydrazine hydrate, 55% (Hydrazine, 35%)
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Indicazioni di pericolo
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Consigli di prudenza
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informazioni supplementari
  • MIXTURE LIST-Contains : Hydrazine

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato