missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrazine hydrate, 80% (Hydrazine, 51%), Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Specifications
| Boiling Point | 117.2°C |
| Melting Point | -57.0°C |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 1 kg |
| Linear Formula | H2NNH2·xH2O |
| Fieser | 01,434; 02,211; 03,153; 04,248; 05,327; 06,280; 07,170; 16,174 |
| Flash Point | 91°C |
| Merck Index | 14, 4771 |
| Solubility Information | Solubility in water: miscible. Other solubilities: miscible with alcohol,insoluble in chloroform and ether |
| Show More |
Safety and Handling
Product Identifier
- Hydrazine hydrate, 80% (Hydrazine, 51%)
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Carcinogenicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H331-Toxic if inhaled.
- H350-May cause cancer.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?